Product Image
Thermo Scientific Chemicals

Nobiletin

CAS: 478-01-3 | C21H22O8 | 402.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 463520010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS478-01-3
IUPAC Name2-(3,4-dimethoxyphenyl)-5,6,7,8-tetramethoxy-4H-chromen-4-one
Molecular FormulaC21H22O8
InChI KeyMRIAQLRQZPPODS-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1OC)C1=CC(=O)C2=C(OC)C(OC)=C(OC)C(OC)=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point136°C to 140°C
Water=<5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only