Product Image
Thermo Scientific Chemicals

Oxaprozin

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 463530050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS21256-18-8
IUPAC Name3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid
Molecular FormulaC18H15NO3
InChI KeyOFPXSFXSNFPTHF-UHFFFAOYSA-N
SMILESOC(=O)CCC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Infrared spectrumConforms
Melting point161°C to 165°C
Loss on drying=<0.3 % (105°C, 2 h)
Appearance (Color)White to light beige
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only