Product Image
Thermo Scientific Chemicals

Nalidixic acid sodium salt

CAS: 3374-05-8 | C12H11N2NaO3 | 254.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 463550050
Price (USD)
92.65
Online exclusive
103.00
Save 10.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
92.65
Online exclusive
103.00
Save 10.35
Each
Chemical Identifiers
CAS3374-05-8
IUPAC Namesodium 1-ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate
Molecular FormulaC12H11N2NaO3
InChI KeyROKRAUFZFDQWLE-UHFFFAOYSA-M
SMILES[Na+].CCN1C=C(C([O-])=O)C(=O)C2=CC=C(C)N=C12
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Water=<7 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only