Product Image
Thermo Scientific Chemicals

Aliquat™ 336TG

CAS: 63393-96-4 | C25H54ClN | 404.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Catalog number 463570025
Price (USD)
-
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS63393-96-4
IUPAC Namemethyltrioctylazanium chloride
Molecular FormulaC25H54ClN
InChI KeyXKBGEWXEAPTVCK-UHFFFAOYSA-M
SMILES[Cl-].CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to amber
Appearance (Form)Viscous liquid
Infrared spectrumConforms
Refractive index1.4655 to 1.4715 (20°C, 589 nm)
Assay112.0 to 117.2 (Quaternary value)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only