Product Image
Thermo Scientific Chemicals

Axitinib

CAS: 319460-85-0 | C22H18N4OS | 386.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 463800010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS319460-85-0
IUPAC NameN-methyl-2-({3-[(1E)-2-(pyridin-2-yl)ethenyl]-1H-indazol-6-yl}sulfanyl)benzamide
Molecular FormulaC22H18N4OS
InChI KeyRITAVMQDGBJQJZ-FMIVXFBMSA-N
SMILESCNC(=O)C1=CC=CC=C1SC1=CC=C2C(NN=C2\C=C\C2=CC=CC=N2)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Infrared spectrumConforms
Loss on drying=<2 %
Appearance (Color)Almost white to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only