Finnpipette™ F3 Variable Volume Single Channel Pipettes
Imagem ilustrativa. O produto pode variar
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Thermo Scientific™
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Support every day pipetting in comfort and with ideal function using these manual variable volume pipettes.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoVolume (Metric)Color
4640060100 to 1000 μLBlue
46400001 to 10 μLPink
46400101 to 10 μLYellow
46400202 to 20 μLTurquoise
46400302 to 20 μLYellow
464004010 to 100 μLYellow
464005020 to 200 μLYellow
46400701 to 10 mLRed
46400805 to 50 μLTurquoise
46400905 to 50 μLYellow
46401000.5 to 5 mLGreen
Número do catálogo 4640060
Preço (BRL)
-
Volume (Metric):
100 to 1000 μL
Color:
Blue
Thermo Scientific™ Finnpipette™ F3 Variable Volume Single Channel Pipettes are comfortable to use, and provide ideal functionality in every day pipetting. The Finnpipette F3 pipette features a wide, color-coded supportive finger rest and ergonomic handle design. Finnpipette F3 pipettes are available in single and multichannel models.
Ergonomic Comfort
  • Wide color-coded supportive finger rest for identification support and ergonomics
  • Designed handle softly contours the shape of your hand
  • Large Ergovisio Display for volume recognition
Ideal Function
  • Durable handle material
  • Double-action pipetting button; rotating upper part prevents accidental volume alteration
  • Advanced volume gearing mechanism for enhanced precision and accuracy
  • Super blow-out for efficient delivery of micro-size drops in 50ul models and below
Compatible with Finntip pipette tips, and other tip models
Especificações
Accuracy±1.5 to 0.5%
Compatible TipsFinntip 1000, Filter 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext, Wide 1000
DescriptionFinnpipette F3 100-1000μL
Increments1μL
ModelF3
Precision0.6 to 0.2%
ColorBlue
Number of Channels1
Product LineFinnpipette™ F3
TypeVariable Volume Pipette
Volume (Metric)100 to 1000 μL
Unit SizeEach