Acetic-d3 acid-d, for NMR, 99.5 atom % D, packaged in 0.75 ml ampoules
Thermo Scientific Chemicals

Acetic-d3 acid-d, for NMR, 99.5 atom % D, packaged in 0.75 ml ampoules

CAS: 1186-52-3 | C2H4O2 | 64.08 g/mol
Have Questions?
Catalog NumberQuantity
4640900757.5 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 464090075
Price (USD)
-
Quantity:
7.5 mL
Request bulk or custom format
Chemical Identifiers
CAS1186-52-3
IUPAC Name(²H₃)ethan(²H)oic acid
Molecular FormulaC2H4O2
InChI KeyQTBSBXVTEAMEQO-GUEYOVJQSA-N
SMILES[2H]OC(=O)C([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only