NHS-Fluorescein (5/6-carboxyfluorescein succinimidyl ester), mixed isomer, 100 mg - FAQs
NHS-Fluorescein (5/6-carboxyfluorescein succinimidyl ester), mixed isomer, 100 mg - FAQs
View additional product information for NHS-Fluorescein (5/6-carboxyfluorescein succinimidyl ester), mixed isomer - FAQs (46410, 46409)
1 product FAQs found
Can you provide the shelf-life for NHS-Fluorescein (5/6-carboxyfluorescein succinimidyl ester), mixed isomer?
NHS-Fluorescein (5/6-carboxyfluorescein succinimidyl ester), mixed isomer is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.