F1-ClipTip™ Variable Volume Single Channel Pipettes
Thermo Scientific™
F1-ClipTip™ Variable Volume Single Channel Pipettes
Lock tips firmly to the pipette with a complete seal with ClipTip technology, so they will not loosen or fall off. Lightweight handle design combined with minimal tip attachment and ejection forces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Compatible Tips
4641210N20 to 200 μLClipTip 200
4641180N2 to 20 μLClipTip 20
4641190N5 to 50 μLClipTip 50
4641200N10 to 100 μLClipTip 200
4641220N30 to 300 μLClipTip 300
4641230N100 to 1000 μLClipTip 1000
4641310N0.1 to 2 μLClipTip 12.5 Ext
4641320N1 to 10 μLClipTip 12.5 Ext
Catalog number 4641210N
Price (USD)
342.75
Special offer
Online exclusive
Ends: 31-Dec-2026
457.00
Save 114.25
Each
Volume (Metric):
20 to 200 μL
Compatible Tips:
ClipTip 200
Price (USD)
342.75
Special offer
Online exclusive
Ends: 31-Dec-2026
457.00
Save 114.25
Each
Ensure a secure fit with ClipTip technology, which locks tips firmly to the pipette with a complete seal so they will not loosen or fall off. Lightweight handle design combined with minimal tip attachment and ejection forces will increase the conform in pipetting. Feel the difference.
Worry-free pipetting is finally here with the only tip that clips.
Sealed in security of ClipTip Technology. Feel the difference.
Eliminate the frustration of banging tips on your pipette
With a light touch, each tip is clipped securely on the pipette.
Reduce the risk of loose or leaking tips
When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles.
Enable consistent and reproducible pipetting from user to user
Higher-quality results and more efficient research. Learn more
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of repetitive strain injuries
Secure Volume Lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Improved small volume pipetting
New 0.1-2 μL model and improved versions 1-10 μL models allow optimized performance in your small volume applications.
Wide Selection of ClipTip Pipette Tips
Compatible sizes with F1-ClipTip pipettes: 12.5 Ext, 20, 50, 200, 300, 300 Ext and 1000μL
Life-time Warranty
With convenient online registration, and may vary by country.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
Specifications
Compatible TipsClipTip 200
DescriptionF1-ClipTip
Increments0.2μL
ColorYellow
Number of Channels1
Product LineF1-ClipTip
TypeVariable Volume Pipette
Volume (Metric)20 to 200 μL
Unit SizeEach