Product Image
Thermo Scientific Chemicals

Carteolol hydrochloride

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 464320010
Price (USD)
136.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
136.00
Each
Chemical Identifiers
CAS51781-21-6
IUPAC Namehydrogen 5-[3-(tert-butylamino)-2-hydroxypropoxy]-1,2,3,4-tetrahydroquinolin-2-one chloride
Molecular FormulaC16H25ClN2O3
InChI KeyFYBXRCFPOTXTJF-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(C)(C)NCC(O)COC1=CC=CC2=C1CCC(=O)N2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=98.5 %
Loss on drying=<0.5 % (1 g, 105°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only