Product Image
Thermo Scientific Chemicals

Bifenox

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 464600010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS42576-02-3
IUPAC Namemethyl 5-(2,4-dichlorophenoxy)-2-nitrobenzoate
Molecular FormulaC14H9Cl2NO5
InChI KeySUSRORUBZHMPCO-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(OC2=CC=C(Cl)C=C2Cl)=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to yellow to light beige
Appearance (Form)Crystalline powder or crystals
Melting point83°C to 89°C
HPLC>=98.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only