Product Image
Thermo Scientific Chemicals

Potassium (trifluoromethyl)trifluoroborate

CAS: 42298-15-7 | CBF6K | 175.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 464730050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS42298-15-7
IUPAC Namepotassium trifluoro(trifluoromethyl)boranuide
Molecular FormulaCBF6K
InChI KeyUOGBGCVSVMQVBR-UHFFFAOYSA-N
SMILES[K+].F[B-](F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or solid
NMR (19F-NMR)>=96 %
Infrared spectrumConforms
Appearance (Color)White to yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only