Product Image
Thermo Scientific Chemicals

Benidipine hydrochloride

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 465100010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS91599-74-5
IUPAC Namehydrogen 3-(3R)-1-benzylpiperidin-3-yl 5-methyl (4R)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate chloride
Molecular FormulaC28H32ClN3O6
InChI KeyKILKDKRQBYMKQX-MIPPOABVSA-N
SMILES[H+].[Cl-].COC(=O)C1=C(C)NC(C)=C([C@@H]1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)O[C@@H]1CCCN(CC2=CC=CC=C2)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only