Product Image
Thermo Scientific Chemicals

Pranlukast hemihydrate

CAS: 150821-03-7 | C54H48N10O9 | 981.04 g/mol
Have Questions?
Catalog NumberQuantity
465331000100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465331000
Price (USD)
-
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS150821-03-7
IUPAC Namebis(N-[4-oxo-2-(2H-1,2,3,4-tetrazol-5-yl)-4H-chromen-8-yl]-4-(4-phenylbutoxy)benzamide) hydrate
Molecular FormulaC54H48N10O9
InChI KeyMSXTUBJFNBZPGC-UHFFFAOYSA-N
SMILESO.O=C(NC1=C2OC(=CC(=O)C2=CC=C1)C1=NNN=N1)C1=CC=C(OCCCCC2=CC=CC=C2)C=C1.O=C(NC1=C2OC(=CC(=O)C2=CC=C1)C1=NNN=N1)C1=CC=C(OCCCCC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point231°C to 234°C
Loss on drying1.5 % to 2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only