Lomustine
Lomustine
Lomustine
Thermo Scientific Chemicals

Lomustine

CAS: 13010-47-4 | C9H16ClN3O2 | 233.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 465492500
Price (USD)
126.65
Online Exclusive
140.00
Save 13.35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
126.65
Online Exclusive
140.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS13010-47-4
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=97.5 %
Water=<1 %
Appearance (Color)Light yellow to yellow
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only