Product Image
Thermo Scientific Chemicals

Pomalidomide

CAS: 19171-19-8 | C13H11N3O4 | 273.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 465700010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS19171-19-8
IUPAC Name4-amino-2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC13H11N3O4
InChI KeyUVSMNLNDYGZFPF-UHFFFAOYNA-N
SMILESNC1=CC=CC2=C1C(=O)N(C1CCC(=O)NC1=O)C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<1 %
HPLC>=98.5 %
Appearance (Form)Powder
Appearance (Color)Yellow to green
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only