Biphenyl-3,3',5,5'-tetracarboxylic acid, Thermo Scientific Chemicals
Biphenyl-3,3',5,5'-tetracarboxylic acid, Thermo Scientific Chemicals
Biphenyl-3,3',5,5'-tetracarboxylic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Biphenyl-3,3',5,5'-tetracarboxylic acid, Thermo Scientific Chemicals

CAS: 4371-28-2 | C16H10O8 | 330.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Catalog number 465831000
Price (USD)
-
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS4371-28-2
IUPAC Name[1,1'-biphenyl]-3,3',5,5'-tetracarboxylic acid
Molecular FormulaC16H10O8
InChI KeyQURGMSIQFRADOZ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=CC(=C1)C(O)=O)C1=CC(=CC(=C1)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)powder or crystalline solid
Infrared spectrumConforms
Titration with NaOH94.0 to 106.0 %
Water=<2.5 %
HPLC>=94.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only