1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea
1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea
1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea
Thermo Scientific Chemicals

1,3-Bis[3,5-bis(trifluoromethyl)phenyl]thiourea

CAS: 1060-92-0 | C17H8F12N2S | 500.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 465880010
Price (USD)
114.65
Special offer
Online exclusive
Ends: 28-Dec-2026
135.00
Save 20.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
114.65
Special offer
Online exclusive
Ends: 28-Dec-2026
135.00
Save 20.35
Each
Chemical Identifiers
CAS1060-92-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only