1,1'-Biadamantane, Thermo Scientific Chemicals
1,1'-Biadamantane, Thermo Scientific Chemicals
1,1'-Biadamantane, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1'-Biadamantane, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 465930010
Price (USD)
196.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
196.00
Each
Chemical Identifiers
CAS3732-31-8
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Melting point285°C to 295°C
GC>=96.0 %
Appearance (Color)White to almost white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only