Glass beads, 5 mm - FAQs
Glass beads, 5 mm - FAQs
View additional product information for Glass beads, 5 mm - FAQs (465955000, 465951000)
2 product FAQs found
What is the Littleton softening point of the Glass beads, 5 mm?
The Glass beads, 5 mm have the following Littleton softening point: TL=670 ± 10 degrees C.
What type of glass are the Glass beads, 5 mm?
The Glass beads, 5 mm are non-acid washed, soda lime glass.