Thiomorpholine 1,1-dioxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thiomorpholine 1,1-dioxide, Thermo Scientific Chemicals

CAS: 39093-93-1 | C4H9NO2S | 135.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466030050
Price (EUR)
136,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
136,00
Each
Chemical Identifiers
CAS39093-93-1
IUPAC Name1λ⁶-thiomorpholine-1,1-dione
Molecular FormulaC4H9NO2S
InChI KeyNDOVLWQBFFJETK-UHFFFAOYSA-N
SMILESO=S1(=O)CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Titration with HClO497.5 to 102.5 %
Infrared spectrumConforms
Melting point68°C to 71°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only