3,3'-Iminobis(N,N-dimethylpropylamine)
Thermo Scientific Chemicals

3,3'-Iminobis(N,N-dimethylpropylamine)

CAS: 6711-48-4 | C10H25N3 | 187.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Catalog number 466340250
Price (USD)
-
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS6711-48-4
IUPAC Name(3-{[3-(dimethylamino)propyl]amino}propyl)dimethylamine
Molecular FormulaC10H25N3
InChI KeyBXYVQNNEFZOBOZ-UHFFFAOYSA-N
SMILESCN(C)CCCNCCCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Total base96.0 to 104.0 %
Appearance (Color)Colorless to brown
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4470 to 1.4510 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only