Product Image
Thermo Scientific Chemicals

5'-Ethynyl-2'-deoxyuridine

252.23 g/mol
Have Questions?
Catalog NumberQuantity
4663800101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466380010
Price (USD)
522.65
Special offer
Online exclusive
Ends: 28-Dec-2026
615.00
Save 92.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
522.65
Special offer
Online exclusive
Ends: 28-Dec-2026
615.00
Save 92.35
Each
Chemical Identifiers
CAS61135-33-9
IUPAC Name5-ethynyl-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC11H12N2O5
InChI KeyCDEURGJCGCHYFH-DJLDLDEBSA-N
SMILESOC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C#C)C(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=96.0 %
Loss on drying=<1.0 %
Appearance (Color)White to slightly yellow
Appearance (Form)powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only