Triptolide - FAQs
Triptolide - FAQs
View additional product information for Triptolide - FAQs (466840250)
2 product FAQs found
What storage conditions should I use for Triptolide?
We recommend storing Triptolide in powder form at -20°C, desiccated. Once in solution, it should be stored at -20°C and used within 1 month to prevent loss of potency. We suggest you make aliquots of the stock solution to avoid multiple freeze/thaw cycles.
How can I prepare a 10 mM stock solution of Triptolide?
To prepare a 10 mM stock of Triptolide, we recommend that you reconstitute 5 mg of powder in 1.38 mL of DMSO.