What storage conditions should I use for Triptolide?
We recommend storing Triptolide in powder form at -20°C, desiccated. Once in solution, it should be stored at -20°C and used within 1 month to prevent loss of potency. We suggest you make aliquots of the stock solution to avoid multiple freeze/thaw cycles.