Dipropylene glycol monobutyl ether
Thermo Scientific Chemicals

Dipropylene glycol monobutyl ether

CAS: 29911-28-2 | C10H22O3 | 190.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Catalog number 467260010
Price (USD)
44.20
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
44.20
Each
Chemical Identifiers
CAS29911-28-2
IUPAC Name1-[(1-butoxypropan-2-yl)oxy]propan-2-ol
Molecular FormulaC10H22O3
InChI KeyCUVLMZNMSPJDON-UHFFFAOYNA-N
SMILESCCCCOCC(C)OCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.4250 to 1.4270 (20°C, nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only