4-Methyltetrahydropyran, stabilized with BHT
Thermo Scientific Chemicals

4-Methyltetrahydropyran, stabilized with BHT

CAS: 4717-96-8 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Catalog number 467320010
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS4717-96-8
IUPAC Name4-methyloxane
Molecular FormulaC6H12O
InChI KeyOVRKATYHWPCGPZ-UHFFFAOYSA-N
SMILESCC1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.4170 to 1.4200 (20°C, 589 nm)
Appearance (Form)liquid
GC>=98.0 %
Stabilizer20 ppm BHT
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only