1,4-Benzenedimethanethiol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Benzenedimethanethiol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 467610050
Price (USD)
419.65
Online exclusive
466.00
Save 46.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
419.65
Online exclusive
466.00
Save 46.35
Each
Chemical Identifiers
CAS105-09-9
IUPAC Name[4-(sulfanylmethyl)phenyl]methanethiol
Molecular FormulaC8H10S2
InChI KeyIYPNRTQAOXLCQW-UHFFFAOYSA-N
SMILESSCC1=CC=C(CS)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point40°C to 45°C
Appearance (Color)White
Appearance (Form)Powder, crystals or solid
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only