2-Phenylindene, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Phenylindene, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 467770050
Price (USD)
313.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
313.00
Each
Chemical Identifiers
CAS4505-48-0
IUPAC Name2-phenyl-1H-indene
Molecular FormulaC15H12
InChI KeyBSBXLZYWGGAVHD-UHFFFAOYSA-N
SMILESC1C2=CC=CC=C2C=C1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or chunks
Melting point165°C to 169°C
Appearance (Color)White to light yellow to light brown
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only