2,7-Dibromocarbazole
Thermo Scientific Chemicals

2,7-Dibromocarbazole

CAS: 136630-39-2 | C12H7Br2N | 325.00 g/mol
Have Questions?
Catalog NumberQuantity
4677800101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 467780010
Price (USD)
65.60
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
65.60
Each
Chemical Identifiers
CAS136630-39-2
IUPAC Name2,7-dibromo-9H-carbazole
Molecular FormulaC12H7Br2N
InChI KeyQPTWWBLGJZWRAV-UHFFFAOYSA-N
SMILESBrC1=CC2=C(C=C1)C1=C(N2)C=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder and or crystals
Melting point230°C to 234°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only