Yeast extract, for biochemistry
Thermo Scientific Chemicals

Yeast extract, for biochemistry

CAS: 8013-01-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 468550010
Price (USD)
210.00
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
210.00
Each
Chemical Identifiers
CAS8013-01-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to beige to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only