Pyrrolo[2,1-f][1,2,4]triazin-4-amine, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyrrolo[2,1-f][1,2,4]triazin-4-amine, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 468990010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS159326-68-8
IUPAC Namepyrrolo[2,1-f][1,2,4]triazin-4-amine
Molecular FormulaC6H6N4
InChI KeyVSPXQZSDPSOPRO-UHFFFAOYSA-N
SMILESNC1=NC=NN2C=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Almost white to grey
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only