2,3-Dicyanopyrazine, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Dicyanopyrazine, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 469000050
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS13481-25-9
IUPAC Namepyrazine-2,3-dicarbonitrile
Molecular FormulaC6H2N4
InChI KeyOTVZGAXESBAAQQ-UHFFFAOYSA-N
SMILESN#CC1=NC=CN=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to brown
Infrared spectrumConforms
Melting point131°C to 135°C
GC>=97.5 %
Appearance (Form)Crystalline powder or crystals and or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only