Sodium thiomethoxide, 90+%, technical
Thermo Scientific Chemicals

Sodium thiomethoxide, 90+%, technical

CAS: 5188-07-8 | CH3NaS | 70.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
Catalog number 469890050
Price (USD)
128.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
128.00
Each
Chemical Identifiers
CAS5188-07-8
IUPAC Namesodium methylsulfanide
Molecular FormulaCH3NaS
InChI KeyRMBAVIFYHOYIFM-UHFFFAOYSA-M
SMILES[Na+].C[S-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light brown
Titration Iodimetric>=90.0 %
Appearance (Form)Powder or granular powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only