You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Industrial Hygiene and Environmental Monitoring
▸
Gas and Vapor Detection
▸
Source Gas Monitors
▸
Model 46i-HL High Level Nitrous Oxide Analyzer
Thermo Scientific™
Model 46i-HL High Level Nitrous Oxide Analyzer
This product has been discontinued. We will offer technical support and service support as allowed by parts availability.
Have Questions?
Catalog Number
Detectable Analytes
46IHL
Nitrous Oxide
Catalog number
46IHL
Price (USD)
-
Detectable Analytes:
Nitrous Oxide
This product has been discontinued. We will offer technical support and service support as allowed by parts availability.
This product has been discontinued. We will offer technical support and service support as allowed by parts availability.
Specifications
Description
High Level Nitrous Oxide Analyzer
Detectable Analytes
Nitrous Oxide
Unit Size
Each