CD3e Monoclonal Antibody (145-2C11), APC-eFluor™ 780, eBioscience™ - FAQs
CD3e Monoclonal Antibody (145-2C11), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD3e Monoclonal Antibody (145-2C11), APC-eFluor™ 780, eBioscience™ - FAQs (47-0031-80, 47-0031-82)