CD3e Monoclonal Antibody (eBio500A2 (500A2)), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD3e Monoclonal Antibody (eBio500A2 (500A2)), APC-eFluor™ 780, eBioscience™ - FAQs (47-0033-82, 47-0033-80)