CD3 Monoclonal Antibody (OKT3), APC-eFluor™ 780, eBioscience™ - FAQs
CD3 Monoclonal Antibody (OKT3), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD3 Monoclonal Antibody (OKT3), APC-eFluor™ 780, eBioscience™ - FAQs (47-0037-42, 47-0037-41)