CD3 Monoclonal Antibody (UCHT1), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD3 Monoclonal Antibody (UCHT1), APC-eFluor™ 780, eBioscience™ - FAQs (47-0038-41, 47-0038-42)