CD4 Monoclonal Antibody (GK1.5), APC-eFluor™ 780, eBioscience™ - FAQs
CD4 Monoclonal Antibody (GK1.5), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (GK1.5), APC-eFluor™ 780, eBioscience™ - FAQs (47-0041-80, 47-0041-82)