CD4 Monoclonal Antibody (RM4-5), APC-eFluor™ 780, eBioscience™ - FAQs
CD4 Monoclonal Antibody (RM4-5), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (RM4-5), APC-eFluor™ 780, eBioscience™ - FAQs (47-0042-82, 47-0042-80)