CD4 Monoclonal Antibody (SK3 (SK-3)), APC-eFluor™ 780, eBioscience™ - FAQs
CD4 Monoclonal Antibody (SK3 (SK-3)), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (SK3 (SK-3)), APC-eFluor™ 780, eBioscience™ - FAQs (47-0047-41, 47-0047-42)