CD4 Monoclonal Antibody (RPA-T4), APC-eFluor™ 780, eBioscience™ - FAQs
CD4 Monoclonal Antibody (RPA-T4), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (RPA-T4), APC-eFluor™ 780, eBioscience™ - FAQs (47-0049-42, 47-0049-41)