CD5 Monoclonal Antibody (53-7.3), APC-eFluor™ 780, eBioscience™ - FAQs
CD5 Monoclonal Antibody (53-7.3), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD5 Monoclonal Antibody (53-7.3), APC-eFluor™ 780, eBioscience™ - FAQs (47-0051-80, 47-0051-82)