CD8a Monoclonal Antibody (53-6.7), APC-eFluor™ 780, eBioscience™ - FAQs
CD8a Monoclonal Antibody (53-6.7), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (53-6.7), APC-eFluor™ 780, eBioscience™ - FAQs (47-0081-80, 47-0081-82)