CD8b Monoclonal Antibody (eBioH35-17.2 (H35-17.2)), APC-eFluor™ 780, eBioscience™ - FAQs
CD8b Monoclonal Antibody (eBioH35-17.2 (H35-17.2)), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD8b Monoclonal Antibody (eBioH35-17.2 (H35-17.2)), APC-eFluor™ 780, eBioscience™ - FAQs (47-0083-82)