CD8a Monoclonal Antibody (RPA-T8), APC-eFluor™ 780, eBioscience™ - FAQs
CD8a Monoclonal Antibody (RPA-T8), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (RPA-T8), APC-eFluor™ 780, eBioscience™ - FAQs (47-0088-41, 47-0088-42)