CD11b Monoclonal Antibody (M1/70), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD11b Monoclonal Antibody (M1/70), APC-eFluor™ 780, eBioscience™ - FAQs (47-0112-82, 47-0112-80)