CD11c Monoclonal Antibody (N418), APC-eFluor™ 780, eBioscience™ - FAQs
CD11c Monoclonal Antibody (N418), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD11c Monoclonal Antibody (N418), APC-eFluor™ 780, eBioscience™ - FAQs (47-0114-80, 47-0114-82)