CD11c Monoclonal Antibody (3.9), APC-eFluor™ 780, eBioscience™ - FAQs
CD11c Monoclonal Antibody (3.9), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD11c Monoclonal Antibody (3.9), APC-eFluor™ 780, eBioscience™ - FAQs (47-0116-42)